Skip to Content
Merck
All Photos(1)

Key Documents

470449

Sigma-Aldrich

Tetrakis[1-[[4-alkyl(C11-C13)phenyl]sulfonyl]-(2S)-pyrrolidinecarboxylate]dirhodium(II)

Synonym(s):

Rh2(S-DOSP)4

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352005
NACRES:
NA.22

form

solid

optical activity

[α]20/D −165°, c = 1 in chloroform

reaction suitability

reaction type: Asymmetric synthesis
reaction type: C-H Activation
reagent type: catalyst
reaction type: Cyclopropanations

mp

195-200 °C (lit.)

storage temp.

2-8°C

Application

Catalyst for asymmetric cyclopropanations.
Catalyst involved in:
  • Stereoselective reactions of aryl- and styryldiazoacetates
  • Enantioselective C-H carbene insertions
  • Asymmetric cyclopropenation of aryl alkynes
  • Intermolecular C-H olefination

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Davies, H.M.L. Hutcheson, D.K.
Tetrahedron Letters, 34, 7243-7243 (1993)
Davies, H.M.L. et al.
Journal of the American Chemical Society, 118, 6897-6897 (1996)
Davies, H.M.L. et al.
Tetrahedron Letters, 38, 1741-1741 (1997)
Davies, H.M.L. et al.
Tetrahedron Letters, 37, 4133-4133 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service