466476
1,3-Bis(methoxycarbonyl)-2-methyl-2-thiopseudourea
97%
About This Item
Recommended Products
Assay
97%
mp
102-105 °C (lit.)
SMILES string
COC(=O)N\C(SC)=N/C(=O)OC
InChI
1S/C6H10N2O4S/c1-11-5(9)7-4(13-3)8-6(10)12-2/h1-3H3,(H,7,8,9,10)
InChI key
KHBXLYPOXVQKJG-UHFFFAOYSA-N
General description
Application
- methyl (6-methyl-4-oxo-4,5-dihydro-3H-pyrrolo[3,2-d]pyrimidin-2-yl)carbamate[1]
- 2-amino-5-benzyl-6-methyl-3,5-dihydro-4H-pyrrolo[3,2-d]pyrimidin-4-one[1]
- methyl-{5-[(3-hydroxypropyl)thio]-1H-benzo[d]imidazol-2-yl}carbamate (hydroxyalbendazole)[2]
- methyl-{5-[(4-hydroxyphenyl)thio]-1H-benzo[d]imidazol-2-yl} carbamate (hydroxyfenbendazole)[2]
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service