Skip to Content
Merck
All Photos(1)

Key Documents

463833

Sigma-Aldrich

Scandium(III) oxalate hydrate

≥99.99% trace metals basis

Synonym(s):

Scandium oxalate (2:3) hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sc2(C2O4)3 · xH2O
CAS Number:
Molecular Weight:
353.97 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

≥99.99% trace metals basis

form

powder

SMILES string

O.O=C1O[Sc](OC1=O)OC(=O)C(=O)O[Sc]2OC(=O)C(=O)O2

InChI

1S/3C2H2O4.H2O.2Sc/c3*3-1(4)2(5)6;;;/h3*(H,3,4)(H,5,6);1H2;;/q;;;;2*+3/p-6

InChI key

UBLURKQDJDHWDF-UHFFFAOYSA-H

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service