Skip to Content
Merck
All Photos(1)

Key Documents

454281

Sigma-Aldrich

2-Fluoro-5-nitrobenzaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(NO2)CHO
CAS Number:
Molecular Weight:
169.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

57-60 °C (lit.)

functional group

aldehyde
fluoro
nitro

SMILES string

[O-][N+](=O)c1ccc(F)c(C=O)c1

InChI

1S/C7H4FNO3/c8-7-2-1-6(9(11)12)3-5(7)4-10/h1-4H

InChI key

VVXFDFQEIRGULC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel synthesis of dibenzo [b,g] 1, 5-oxazocines.
Ouyang X and Kiselyov AS.
Tetrahedron Letters, 40(32), 5827-5830 (1999)
J Craig Ruble et al.
Journal of the American Chemical Society, 131(11), 3991-3997 (2009-03-06)
PNU-286607 is the first member of a promising, novel class of antibacterial agents that act by inhibiting bacterial DNA gyrase, a target of clinical significance. Importantly, PNU-286607 displays little cross-resistance with marketed antibacterial agents and is active against methicillin-resistant staphylococcus
The preparation of some polymethine astrazon dyes.
Gale DJ and Wilshire JFK.
Australian Journal of Chemistry, 23(5), 1063-1068 (1970)
A valuable heterocyclic ring transformation: from isoxazolin-5 (2H)-ones to quinolines.
Abbiati G, et al.
Tetrahedron, 59(50), 9887-9893 (2003)
Facile solid phase synthesis of 1, 2-disubstituted-6-nitro-1, 4-dihydroquinazolines using a tetrafunctional scaffold.
Wang X MLA, et al.
Tetrahedron Letters, 46(3), 427-430 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service