Recommended Products
Assay
98%
refractive index
n20/D 1.421 (lit.)
bp
120 °C/50 mmHg (lit.)
density
0.811 g/mL at 25 °C (lit.)
SMILES string
CC(C)(C)[Si](C)(C)OCC=C
InChI
1S/C9H20OSi/c1-7-8-10-11(5,6)9(2,3)4/h7H,1,8H2,2-6H3
InChI key
DHWVPYLVNAKSEU-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
91.4 °F - closed cup
Flash Point(C)
33 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The Journal of organic chemistry, 78(9), 4231-4238 (2013-04-16)
Twelve chiral bis-3,4-diazaphospholane ligands and six alkene substrates (styrene, vinyl acetate, allyloxy-tert-butyldimethylsilane, (E)-1-phenyl-1,3-butadiene, 2,3-dihydrofuran, and 2,5-dihydrofuran) probe the influence of steric bulk on the activity and selectivity of asymmetric hydroformylation (AHF) catalysts. Reaction of an enantiopure bisdiazaphospholane tetraacyl fluoride with
Poly (ferrocenylsilane-b lock-methacrylates) via Sequential Anionic and Atom Transfer Radical Polymerization.
Macromolecules, 37(5), 1686-1690 (2004)
European journal of immunology, 50(10), 1537-1549 (2020-05-12)
The IL-7 receptor specific α chain, CD127, can be expressed both as a membrane-associated (mCD127) and a soluble form (sCD127), however, the mechanisms involved in their regulation remain to be defined. We first demonstrated in primary human CD8+ T cells
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service