Skip to Content
Merck
All Photos(1)

Key Documents

436380

Sigma-Aldrich

Tetraamminepalladium(II) acetate

Synonym(s):

Tetraamminepalladium(2+) diacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pd(NH3)4(CH3CO2)2
CAS Number:
Molecular Weight:
292.63
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

solid

reaction suitability

core: aluminum
reagent type: catalyst

mp

183-186 °C (dec.) (lit.)

SMILES string

N.N.N.N.O.O.CC(=O)O[Pd]OC(C)=O

InChI

1S/2C2H4O2.4H3N.2H2O.Pd/c2*1-2(3)4;;;;;;;/h2*1H3,(H,3,4);4*1H3;2*1H2;/q;;;;;;;;+2/p-2

InChI key

TXKHLAUDWHNZBC-UHFFFAOYSA-L

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J P Berry
Journal of submicroscopic cytology, 19(1), 53-56 (1987-01-01)
Palladium salt has been used for some time in experimental therapy protocols; with this in mind, we carried out a study of the effect of tetramine dichloro-palladium (soluble salt) upon kidney cells. Using an electron microprobe, we were able to
J Ma
The Journal of general physiology, 102(6), 1031-1056 (1993-12-01)
The effects of ruthenium red and the related compounds tetraamine palladium (4APd) and tetraamine platinum (4APt) were studied on the ryanodine activated Ca2+ release channel reconstituted in planar bilayers with the immunoaffinity purified ryanodine receptor. Ruthenium red, applied at submicromolar

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service