All Photos(1)
About This Item
Linear Formula:
Cl2C6H2-1,2-(CONH2)2
CAS Number:
Molecular Weight:
233.05
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Assay
97%
mp
240 °C (dec.) (lit.)
SMILES string
NC(=O)c1cc(Cl)c(Cl)cc1C(N)=O
InChI
1S/C8H6Cl2N2O2/c9-5-1-3(7(11)13)4(8(12)14)2-6(5)10/h1-2H,(H2,11,13)(H2,12,14)
InChI key
KGIXKEAUEDTQOE-UHFFFAOYSA-N
General description
4,5-Dichlorophthalamide (4,5-dichloro-1,2-benzenedicarboxamide) is a phthalamide derivative. It has been synthesized from 5,6-dichloro-1H-isoindole-1,3(2H)-dione and characterized by 1H ,13C-NMR and IR spectra.
Application
4,5-Dichlorophthalamide (4,5-dichloro-1,2-benzenedicarboxamide) may be used as a starting material in the synthesis of 4,5-dichloro-1,2-dicyanobenzene.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Synthesis of water soluble PEGylated (copper) phthalocyanines via Mitsunobu reaction and Cu (i)-catalysed azide-alkyne cycloaddition (CuAAC) click chemistry.
Li M, et al.
Polym. Chem., 4(16), 4405-4411 (2013)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service