Skip to Content
Merck
All Photos(1)

Documents

375012

Sigma-Aldrich

Poly(lauryl methacrylate-co-ethylene glycol dimethacrylate)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

mp

>200 °C (lit.)

SMILES string

CC(=C)C(=O)OCCOC(=O)C(C)=C.CCCCCCCCCCCCOC(=O)C(C)=C

InChI

1S/C16H30O2.C10H14O4/c1-4-5-6-7-8-9-10-11-12-13-14-18-16(17)15(2)3;1-7(2)9(11)13-5-6-14-10(12)8(3)4/h2,4-14H2,1,3H3;1,3,5-6H2,2,4H3

InChI key

YVUPJRVTTNZUEI-UHFFFAOYSA-N

Application

Absorbs and removes oils from surfaces.

Features and Benefits

Absorbs up to four times its weight in lipophilic materials. Minimizes syneresis.
Porous, highly cross-linked copolymer. High oil absorption. 200-1,200μm aggregates of <1μm-sized particles

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service