Skip to Content
Merck
All Photos(1)

Documents

357545

Sigma-Aldrich

Nickel

foil, thickness 1.0 mm, 99.98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ni
CAS Number:
Molecular Weight:
58.69
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

99.98%

form

foil

resistivity

6.97 μΩ-cm, 20°C

thickness

1.0 mm

bp

2732 °C (lit.)

mp

1453 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Ni]

InChI

1S/Ni

InChI key

PXHVJJICTQNCMI-UHFFFAOYSA-N

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ornella Valenti et al.
Cerebral cortex (New York, N.Y. : 1991), 19(3), 658-674 (2008-07-18)
The prefrontal cortex receives multiple inputs from the hippocampal complex, which are thought to drive memory-guided behavior. Moreover, dysfunctions of both regions have been repeatedly associated with several psychiatric disorders. Therefore, understanding the interconnections and modulatory interactions between these regions
A J Feilzer et al.
Nederlands tijdschrift voor tandheelkunde, 120(6), 335-341 (2013-07-19)
Orally applied metal alloys can cause undesirable physical effects. A distinction needs to be made in this respect between local and systemic reactions and toxic and immunological reactions. A case is presented which illustrates this problem. In this case, the
Marilia O S Seijo et al.
Journal of dental education, 77(5), 648-655 (2013-05-10)
Including students' perceptions in the educational process is considered a key component in monitoring the quality of academic programs. This study aimed to evaluate the concept of one's learning experience in endodontic teaching from the perspective of a group of
Palatally anchored maxillary molar mesialization using the mesialslider.
Benedict Wilmes et al.
Journal of clinical orthodontics : JCO, 47(3), 172-179 (2013-05-11)
Fariborz Amini et al.
International journal of orthodontics (Milwaukee, Wis.), 24(1), 43-44 (2013-06-05)
In this report, we present an uncomplicated approach to overcome lingual rolling of mandibular molars during orthodontic treatment, by using rectangular NiTi arch wire in twist fashion through the molar tube. The sequential steps of the twist manoeuvre on a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service