Skip to Content
Merck
All Photos(1)

Key Documents

346071

Sigma-Aldrich

2-Chloro-4-hydroxybenzonitrile

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(OH)CN
CAS Number:
Molecular Weight:
153.57
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

160 °C (dec.) (lit.)

functional group

chloro
nitrile

SMILES string

Oc1ccc(C#N)c(Cl)c1

InChI

1S/C7H4ClNO/c8-7-3-6(10)2-1-5(7)4-9/h1-3,10H

InChI key

BDDVAWDNVWLHDQ-UHFFFAOYSA-N

Application

2-Chloro-4-hydroxybenzonitrile may be used in the synthesis of 6-aminophenanthridines via Suzuki-Miyaura coupling reaction. It may be used in the preparation of 5-bromo-2-chloro-4-hydroxybenzonitrile.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An expeditious synthesis of 6-aminophenanthridines.
Gug F, et al.
Tetrahedron Letters, 46(21), 3725-3727 (2005)
Min Ju Kim et al.
Bioorganic & medicinal chemistry, 19(18), 5468-5479 (2011-08-27)
Novel macrocyclic C-aryl glucoside SGLT2 inhibitors were designed and synthesized. Two different synthetic routes of macrocyclization were adopted to prepare novel ansa SGLT2 inhibitors. Among the compounds tested, [1,7]dioxacyclopentadecine macrocycles possessing methylthiophenyl at the distal ring 40 or ethoxyphenyl at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service