Skip to Content
Merck
All Photos(1)

Documents

333573

Sigma-Aldrich

1-Methylanthracene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12
CAS Number:
Molecular Weight:
192.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

bp

363 °C (lit.)

mp

86-88 °C (lit.)

SMILES string

Cc1cccc2cc3ccccc3cc12

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patricia K Tithof et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 16(11), 1463-1464 (2002-09-03)
Smoking is a major risk factor for endothelial cell injury and subsequent coronary artery disease. Epidemiological studies implicate the phospholipase A2/arachidonic acid cascade in the mechanism by which smoking causes heart disease. However, specific components of cigarette smoke that activate
Mei-Hui Tai et al.
International journal of cancer, 120(9), 1855-1862 (2007-02-03)
Smoking is a well-documented risk factor for the development of pancreatic adenocarcinoma. Although the most abundant polycyclic aromatic hydrocarbons (PAHs) in cigarette smoke are methylated anthracenes and phenanthrenes, the epigenetic toxicity of these compounds has not been extensively studied. We
Capillary gas chromatography/pulsed supersonic jet/fluorescence excitation spectroscopy for the identification of methylanthracenes in a complex environmental sample.
B V Pepich et al.
Analytical chemistry, 58(13), 2825-2830 (1986-11-01)
S R Langhoff et al.
The journal of physical chemistry. A, 102(9), 1632-1646 (2001-09-07)
Calculations are carried out using density functional theory (DFT) to determine the harmonic frequencies and intensities of 1-methylanthracene, 9-methylanthracene, 9-cyanoanthracene, 2-aminoanthracene, acridine, and their positive ions. The theoretical data are compared with matrix-isolation spectra for these species also reported in
Brad L Upham et al.
Cancer science, 99(4), 696-705 (2008-04-02)
Inhibition of gap junctional intercellular communication (GJIC) and the activation of intracellular mitogenic pathways are common hallmarks of epithelial derived cancer cells. We previously determined that the 1-methyl and not the 2-methyl isomer of anthracene, which are prominent cigarette smoke

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service