Skip to Content
Merck
All Photos(1)

Key Documents

309893

Sigma-Aldrich

Diethyl isopropylmalonate

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH(COOC2H5)2
CAS Number:
Molecular Weight:
202.25
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥98%

refractive index

n20/D 1.42 (lit.)

bp

62 °C/0.35 mmHg (lit.)

density

0.981 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C(C(C)C)C(=O)OCC

InChI

1S/C10H18O4/c1-5-13-9(11)8(7(3)4)10(12)14-6-2/h7-8H,5-6H2,1-4H3

InChI key

BYQFBFWERHXONI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethyl isopropylmalonate reacts with chalcone by Michael addition, under high pressure and fluoride catalyst, to form compounds having quarternary C atoms.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High pressure Michael addition catalyzed by fluoride ions.
Angewandte Chemie (International Edition in English), 20(9), 770-771 (1981)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service