284874
5-Nitro-2-furaldehyde semicarbazone
97%
Synonym(s):
5-Nitro-2-furfural semicarbazone, Furacin, NFZ, Nitrofurazone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H6N4O4
CAS Number:
Molecular Weight:
198.14
Beilstein:
86403
EC Number:
MDL number:
UNSPSC Code:
12352100
Assay
97%
mp
242-244 °C (lit.)
SMILES string
NC(=O)N\N=C\c1ccc(o1)[N+]([O-])=O
InChI
1S/C6H6N4O4/c7-6(11)9-8-3-4-1-2-5(14-4)10(12)13/h1-3H,(H3,7,9,11)/b8-3+
InChI key
IAIWVQXQOWNYOU-FPYGCLRLSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service