284580
3-Aminoadipic acid
98%
Synonym(s):
3-Aminohexanedioic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HO2CCH2CH2CH(NH2)CH2CO2H
CAS Number:
Molecular Weight:
161.16
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Assay
98%
mp
205 °C (dec.) (lit.)
SMILES string
NC(CCC(O)=O)CC(O)=O
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service