Skip to Content
Merck
All Photos(1)

Key Documents

276308

Sigma-Aldrich

2-Chloro-1,3-dimethylbenzene

97%

Synonym(s):

2-Chloro-m-xylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3Cl
CAS Number:
Molecular Weight:
140.61
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

form

liquid

refractive index

n20/D 1.527 (lit.)

bp

182-183 °C (lit.)

mp

−36 °C (lit.)

density

1.061 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

Cc1cccc(C)c1Cl

InChI

1S/C8H9Cl/c1-6-4-3-5-7(2)8(6)9/h3-5H,1-2H3

InChI key

VDXLAYAQGYCQEO-UHFFFAOYSA-N

General description

Suzuki coupling reaction of 2-chloro-1,3-dimethylbenzene with arylboronic acids has been investigated[1]. 2-Chloro-1,3-dimethylbenzene undergoes palladium-catalyzed cyanation reaction to afford 2,6-dimethyl benzonitrile[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A practical heterogeneous catalyst for the Suzuki, Sonogashira, and Stille coupling reactions of unreactive aryl chlorides.
Myung-Jong Jin et al.
Angewandte Chemie (International ed. in English), 49(6), 1119-1122 (2010-01-01)
A new palladium catalyst system for the cyanation of aryl chlorides with K4[Fe(CN)6].
Schareina T, et al.
Tetrahedron Letters, 48(7), 1087-1090 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service