Skip to Content
Merck
All Photos(1)

Key Documents

273449

Sigma-Aldrich

(4R)-(−)-2-Thioxo-4-thiazolidinecarboxylic acid

99%

Synonym(s):

(R)-(−)-2-Thioxo-4-thiazolidinecarboxylic acid, Raphanusamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5NO2S2
CAS Number:
Molecular Weight:
163.22
Beilstein:
82632
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

form

powder

optical activity

[α]29/D −86°, c = 2.5 in 0.5 M HCl

reaction suitability

reaction type: solution phase peptide synthesis

mp

179-181 °C (lit.)

application(s)

peptide synthesis

SMILES string

OC(=O)[C@@H]1CSC(=S)N1

InChI

1S/C4H5NO2S2/c6-3(7)2-1-9-4(8)5-2/h2H,1H2,(H,5,8)(H,6,7)/t2-/m0/s1

InChI key

SQUOCHQOQMZGQP-REOHCLBHSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pawel Bednarek et al.
Science (New York, N.Y.), 323(5910), 101-106 (2008-12-20)
Selection pressure exerted by insects and microorganisms shapes the diversity of plant secondary metabolites. We identified a metabolic pathway for glucosinolates, known insect deterrents, that differs from the pathway activated by chewing insects. This pathway is active in living plant
V Amarnath et al.
Chemical research in toxicology, 14(9), 1277-1283 (2001-09-18)
A new method is reported for the analysis of 2-thioxothiazolidine-4-carboxylic acid (TTCA) in urine that is amenable to automation and provides greatly simplified chromatograms. The method comprises the addition of tetrahydro-2-thioxo-2H-1,3-thiazine-4-carboxylic acid, which is chemically similar to TTCA, as internal
S E Sparks et al.
Journal of biochemical and molecular toxicology, 14(1), 26-32 (1999-11-24)
Chloropicrin (CCl3NO2) is a widely used soil fumigant with an unknown mechanism of acute toxicity. We investigated the possible involvement of dechlorination in CCl3NO2 toxicity by considering its metabolism, inhibition of pyruvate and succinate dehydrogenases, cytotoxicity in cultured cells, and
A J Dobson et al.
Acta crystallographica. Section C, Crystal structure communications, 54 ( Pt 11), 1634-1637 (1998-12-19)
The title acid, C4H5NO2S2, crystallized in the noncentrosymmetric space group P2(1)2(1)2(1) with one molecule as the asymmetric unit. Two hydrogen bonds occur, namely, N--H...O, with a donor-acceptor distance of 2.850 (2) A, and O-H...S (resonance induced), with a donor-acceptor distance
Verena Jeschke et al.
Frontiers in plant science, 10, 1560-1560 (2019-12-24)
Dynamically changing environmental conditions promote a complex regulation of plant metabolism and balanced resource investments to development and defense. Plants of the Brassicales order constitutively allocate carbon, nitrogen, and sulfur to synthesize glucosinolates as their primary defense metabolites. Previous findings

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service