Skip to Content
Merck
All Photos(1)

Key Documents

244295

Sigma-Aldrich

2,2-Pentamethylene-1,3-dioxolane

97%

Synonym(s):

1,4-Dioxaspiro[4.5]decane, Cyclohexanone ethylene ketal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14O2
CAS Number:
Molecular Weight:
142.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.459 (lit.)

bp

73 °C/16 mmHg (lit.)

density

1.028 g/mL at 25 °C (lit.)

SMILES string

C1CCC2(CC1)OCCO2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

156.2 °F - closed cup

Flash Point(C)

69 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service