Skip to Content
Merck
All Photos(1)

Key Documents

221104

Sigma-Aldrich

N-(4-Nitrobenzoyl)-L-glutamic acid diethyl ester

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5O2CCH2CH2CH(NHCOC6H4NO2)CO2C2H5
CAS Number:
Molecular Weight:
352.34
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

98%

optical activity

[α]24/D +20.5°, c = 2 in chloroform

mp

92-94 °C (lit.)

application(s)

peptide synthesis

SMILES string

CCOC(=O)CC[C@H](NC(=O)c1ccc(cc1)[N+]([O-])=O)C(=O)OCC

InChI

1S/C16H20N2O7/c1-3-24-14(19)10-9-13(16(21)25-4-2)17-15(20)11-5-7-12(8-6-11)18(22)23/h5-8,13H,3-4,9-10H2,1-2H3,(H,17,20)/t13-/m0/s1

InChI key

OTQQWHKIMACEHP-ZDUSSCGKSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service