Skip to Content
Merck
All Photos(1)

Key Documents

209120

Sigma-Aldrich

Ethyl trimethylsilylacetate

≥98%

Synonym(s):

ETSA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiCH2CO2C2H5
CAS Number:
Molecular Weight:
160.29
Beilstein:
1755902
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥98%

form

liquid

refractive index

n20/D 1.415 (lit.)

bp

156-159 °C (lit.)

density

0.876 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C[Si](C)(C)C

InChI

1S/C7H16O2Si/c1-5-9-7(8)6-10(2,3)4/h5-6H2,1-4H3

InChI key

QQFBQBDINHJDMN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Reaction of ethyl trimethylsilylacetate (ETSA) with dilute hydrochloric acid, dilute alkali, anhydrous HCl, anhydrous bromine and absolute ethanol has been reported[1]. ETSA undergoes condensation with aromatic aldehydes in the presence of base catalyst to yield β-trimethylsiloxycarboxylates[2]. Lithium ETSA reacts with ketones to yield α,β-unsaturated esters[3].

Application

Ethyl trimethylsilylacetate (ETSA) was used in the synthesis of ethyl-2,2-dibromo-2-(trimethylsilyl)acetate[4]. It was also used to silylate the enolizable aldehydes and ketones[5]

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

<73.4 °F - closed cup

Flash Point(C)

< 23 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and Cleavage of Ethyl Trimethylsilylacetate1, 2.
Gold JR, et al.
Journal of the American Chemical Society, 70(9), 2874-2876 (1948)
Synthetic Communications, 19, 2441-2441 (1989)
New synthesis of. alpha.,. beta.-unsaturated carboxylic esters.
Shimoji K, et al.
Journal of the American Chemical Society, 96(5), 1620-1621 (1974)
Mitsuru Shindo et al.
Chemical & pharmaceutical bulletin, 51(4), 477-478 (2003-04-04)
Aliphatic alpha,alpha-dibromo esters, precursors of ynolates, were synthesized via bromination of lithium alpha-bromo ester enolates with 1,2-dibromotetrafluoroethane in good yields. alpha-Trimethylsilyl-alpha,alpha-dibromo esters were synthesized via radical bromination.
Journal of the Chemical Society. Perkin Transactions 1, 3217-3217 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service