Skip to Content
Merck
All Photos(1)

Documents

206571

Sigma-Aldrich

6-Methoxy-8-nitroquinoline

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8N2O3
CAS Number:
Molecular Weight:
204.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

mp

158-160 °C (lit.)

SMILES string

COc1cc([N+]([O-])=O)c2ncccc2c1

InChI

1S/C10H8N2O3/c1-15-8-5-7-3-2-4-11-10(7)9(6-8)12(13)14/h2-6H,1H3

InChI key

MIMUSZHMZBJBPO-UHFFFAOYSA-N

Application

6-Methoxy-8-nitroquinoline was used in the synthesis of:
  • 6-methoxy-8-aminoquinoline, metabolite of primaquine
  • 6-methoxy-8-hydroxylaminoquinoline
  • 2-substituted/2,5-disubstituted-6-methoxy-8-nitroquinolines

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suryanarayana Vangapandu et al.
Bioorganic & medicinal chemistry, 12(10), 2501-2508 (2004-04-28)
We report in vitro antimycobacterial properties of ring-substituted quinolines (series 1-4) constituting 56 analogues against drug-sensitive and drug-resistant M. tuberculosis H37Rv strains. The most effective compounds 2h (R1 = R2 = c-C6H11, R3 = NO2, series 1) and 13g (R1
L J Bolchoz et al.
The Journal of pharmacology and experimental therapeutics, 297(2), 509-515 (2001-04-17)
Primaquine is an important antimalarial agent because of its activity against exoerythrocytic forms of Plasmodium spp. However, methemoglobinemia and hemolytic anemia are dose-limiting side effects of primaquine therapy that limit its efficacy. These hemotoxicities are thought to be mediated by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service