Skip to Content
Merck
All Photos(1)

Documents

193836

Sigma-Aldrich

3-Methoxy-4-nitrobenzyl alcohol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H3(NO2)CH2OH
CAS Number:
Molecular Weight:
183.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

95-97 °C (lit.)

SMILES string

COc1cc(CO)ccc1[N+]([O-])=O

InChI

1S/C8H9NO4/c1-13-8-4-6(5-10)2-3-7(8)9(11)12/h2-4,10H,5H2,1H3

InChI key

AADYWCBPJZAJNU-UHFFFAOYSA-N

Application

3-Methoxy-4-nitrobenzyl alcohol was used in the synthesis of:
  • 3-methoxy-4-nitrobenzyl azide
  • new aryl azides, (Z)-1-(3′-azido-4′-methoxyphenyl)-2-(3″,4″,5″-trimethoxyphenyl)ethane and (Z)-1-(4′-azido-3′-methoxyphenyl)-2-(3″,4″,5″-trimethoxyphenyl)ethane, potentially useful photoaffinity labeling reagents for tubulin
  • 3-methoxy-4-nitrobenzaldehyde via oxidation
  • 4-[3-(2-methylphenyl)ureido]-3-methoxybenzylchloride, required for syntheses of trifluoromethyl-substituted hydantoins

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K G Pinney et al.
Bioorganic & medicinal chemistry, 8(10), 2417-2425 (2000-11-01)
Two new aryl azides, (Z)-1-(3'-azido-4'-methoxyphenyl)-2-(3",4",5"-trimethoxyphenyl)ethene 9 and (Z)-1-(4'-azido-3'-methoxyphenyl)-2-(3",4",5"-trimethoxyphenyl)ethene 5, modeled after the potent antitumor, antimitotic agent combretastatin A-4 (CA-4), have been prepared by chemical synthesis as potentially useful photoaffinity labeling reagents for the colchicine site on beta-tubulin. Aryl azide 9
Jeewoo Lee et al.
Journal of medicinal chemistry, 46(14), 3116-3126 (2003-06-27)
Isosteric replacement of the phenolic hydroxyl group in potent vanilloid receptor (VR1) agonists with the alkylsulfonamido group provides a series of compounds which are effective antagonists to the action of the capsaicin on rat VR1 heterologously expressed in Chinese hamster
Trifluoromethyl-substituted hydantoins, versatile building blocks for rational drug design.
Wehner V, et al.
Tetrahedron, 60(19), 4295-4302 (2004)
Can we predict reactivity for aromatic nucleophilic substitution with [18F] fluoride ion?
Rengan R, et al.
Journal of Labelled Compounds & Radiopharmaceuticals, 33(7), 563-572 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service