Skip to Content
Merck
All Photos(2)

Key Documents

161772

Sigma-Aldrich

β-Methoxystyrene, mixture of cis and trans

technical grade, 90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CHOCH3
CAS Number:
Molecular Weight:
134.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

grade

technical grade

Assay

90%

form

liquid

refractive index

n20/D 1.565 (lit.)

bp

50-56 °C/0.6 mmHg (lit.)

density

1.001 g/mL at 25 °C (lit.)

functional group

ether
phenyl

storage temp.

2-8°C

SMILES string

COC=Cc1ccccc1

InChI

1S/C9H10O/c1-10-8-7-9-5-3-2-4-6-9/h2-8H,1H3

InChI key

CTHJQRHPNQEPAB-UHFFFAOYSA-N

General description

β-Methoxystyrene undergoes reduction in the presence of calcium in liquid ammonia to yield methyl phenethyl ether[1].

Application

β-Methoxystyrene was used in the synthesis of trans, trans-1-(aminomethyl)-2-methoxy-3-phenylcyclopropane[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Calcium metal in liquid ammonia for selective reduction of organic compounds.
Hwu JH, et al.
The Journal of Organic Chemistry, 61(4), 1493-1499 (1996)
Xingliang Lu et al.
The Journal of organic chemistry, 61(25), 8961-8966 (1996-12-13)
trans,trans-1-(Aminomethyl)-2-methoxy-3-phenylcyclopropane (3) was synthesized in three steps from (Z)-beta-methoxystyrene and ethyl diazoacetate. Compound 3 was shown to be a substrate and inactivator of mitochondrial beef liver monoamine oxidase (MAO) with a partition ratio of 1428. MAO-catalyzed oxidation of 3 produces

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service