Skip to Content
Merck
All Photos(1)

Key Documents

161446

Sigma-Aldrich

Methyl 5-chloro-2-methoxybenzoate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(OCH3)CO2CH3
CAS Number:
Molecular Weight:
200.62
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

refractive index

n20/D 1.5466 (lit.)

bp

235-240 °C (lit.)

density

1.259 g/mL at 25 °C (lit.)

SMILES string

COC(=O)c1cc(Cl)ccc1OC

InChI

1S/C9H9ClO3/c1-12-8-4-3-6(10)5-7(8)9(11)13-2/h3-5H,1-2H3

InChI key

HPTHYBXMNNGQEF-UHFFFAOYSA-N

Application

Methyl 5-chloro-2-methoxybenzoate may be used in chemical synthesis.

Biochem/physiol Actions

Methyl 5-chloro-2-methoxybenzoate inhibits copulatory behaviour in male Ixodes ricinus (L.1758).

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Methyl 3-chloro-4-methoxybenzoate, a new candidate semiochemical inhibiting copulation behaviour of Ixodes ricinus (L.) males.
Bouman EAP, et al.
Physiological Entomology, 28(4), 276-282 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service