Skip to Content
Merck
All Photos(2)

Documents

156906

Sigma-Aldrich

2,6-Dimethylbenzoic acid

97%

Synonym(s):

vic.-m-Xylylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3CO2H
CAS Number:
Molecular Weight:
150.17
Beilstein:
1863791
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

114-116 °C (lit.)

SMILES string

Cc1cccc(C)c1C(O)=O

InChI

1S/C9H10O2/c1-6-4-3-5-7(2)8(6)9(10)11/h3-5H,1-2H3,(H,10,11)

InChI key

HCBHQDKBSKYGCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Crystal structure of 2,6-dimethylbenzoic acid was studied by three-dimensional X-ray methods.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The crystal structure of 2, 6-dimethylbenzoic acid.
Anca R, et al.
Acta Crystallographica Section E, Structure Reports Online, 23(6), 1010-1016 (1967)
Marcin Poreba et al.
Cell death and differentiation, 26(2), 229-244 (2018-05-12)
Apical caspases initiate and effector caspases execute apoptosis. Reagents that can distinguish between caspases, particularly apical caspases-8, 9, and 10 are scarce and generally nonspecific. Based upon a previously described large-scale screen of peptide-based caspase substrates termed HyCoSuL, we sought

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service