Skip to Content
Merck
All Photos(1)

Key Documents

155330

Sigma-Aldrich

Cyclodecane

≥90%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20
CAS Number:
Molecular Weight:
140.27
Beilstein:
1900637
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥90%

form

liquid

refractive index

n20/D 1.471 (lit.)

bp

201 °C (lit.)

mp

9-10 °C (lit.)

density

0.871 g/mL at 25 °C (lit.)

SMILES string

C1CCCCCCCCC1

InChI

1S/C10H20/c1-2-4-6-8-10-9-7-5-3-1/h1-10H2

InChI key

LMGZGXSXHCMSAA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The concentration-dependent diffusion rate of cyclodecane has been studied[1].

Application

Cyclodecane was used in the hydrolysis of GM1[2].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Wang et al.
Langmuir : the ACS journal of surfaces and colloids, 29(9), 2935-2945 (2013-01-31)
The structure of a molecule and its concentration can strongly influence diffusional properties for transport in nanoporous materials. We study mass transfer of alkanes in BPL activated carbon using the concentration-swing frequency response method, which can easily discriminate among mass
T Kurita et al.
Journal of lipid research, 41(5), 846-851 (2000-04-29)
Lysoglycosphingolipids were produced from glycosphingolipids by using sphingolipid ceramide N-deacylase, which cleaves the N-acyl linkage between fatty acids and sphingosine bases in various glycosphingolipids. The enzyme reaction was done in a biphasic media prepared with water;-immiscible organic solvent and aqueous

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service