Skip to Content
Merck
All Photos(1)

Documents

141569

Sigma-Aldrich

1-(4-Chlorophenyl)-1-cyclopentanecarboxylic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4C5H8CO2H
CAS Number:
Molecular Weight:
224.68
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

form

solid

mp

162-164 °C (lit.)

SMILES string

OC(=O)C1(CCCC1)c2ccc(Cl)cc2

InChI

1S/C12H13ClO2/c13-10-5-3-9(4-6-10)12(11(14)15)7-1-2-8-12/h3-6H,1-2,7-8H2,(H,14,15)

InChI key

QJNFJEMGWIQMJT-UHFFFAOYSA-N

Biochem/physiol Actions

1-(4-Chlorophenyl)-1-cyclopentanecarboxylic acid reacts with diorganotin(IV) oxide or dichloride to yield organotin(IV) complexes having antitumor activities against various human cancer cell lines.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xianmei Shang et al.
Inorganic chemistry, 50(17), 8158-8167 (2011-07-29)
The organotin(IV) compounds [Me(2)Sn(L)(2)] (1), [Et(2)Sn(L)(2)] (2), [(n)Bu(2)Sn(L)(2)] (3), [(n)Oct(2)Sn(L)(2)] (4), [Ph(2)Sn(L)(2)] (5), and [PhOSnL](6) (6) have been synthesized from the reactions of 1-(4-chlorophenyl)-1-cyclopentanecarboxylic acid (HL) with the corresponding diorganotin(IV) oxide or dichloride. They were characterized by IR and multinuclear

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service