Skip to Content
Merck
All Photos(1)

Key Documents

117803

Sigma-Aldrich

exo-2,3-Epoxynorbornane

97%

Synonym(s):

ENB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10O
CAS Number:
Molecular Weight:
110.15
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

97%

form

solid

mp

122-124 °C (lit.)

SMILES string

C1C[C@H]2C[C@@H]1[C@@H]3O[C@H]23

InChI

1S/C7H10O/c1-2-5-3-4(1)6-7(5)8-6/h4-7H,1-3H2/t4-,5+,6+,7-

InChI key

OHNNZOOGWXZCPZ-RNGGSSJXSA-N

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Noelia Pallarés et al.
Journal of agricultural and food chemistry, 65(47), 10282-10289 (2017-10-27)
The aim of the present study was to develop a multimycotoxin liquid chromatography tandem mass spectrometry (LC-MS/MS) method with a dispersive liquid-liquid microextraction procedure (DLLME) for the analysis of AFs, 3aDON, 15aDON, NIV, HT-2, T-2, ZEA, OTA, ENNs, and BEA
Andrea Šlampová et al.
Analytica chimica acta, 950, 49-56 (2016-12-06)
A tailor-made liquid membrane consisting of a resistive organic solvent (nitrobenzene, NB) and a highly selective non-ionic macrocyclic compound (bambus[6]uril, BU6) was employed for electromembrane extraction (EME) of inorganic anions. BU6 facilitates strong host-guest interactions of its internal cavity with
Masaaki Sato et al.
General thoracic and cardiovascular surgery, 67(12), 1062-1069 (2019-05-18)
Conventional virtual-assisted lung mapping (VAL-MAP), also termed multi-spot preoperative bronchoscopic lung marking, necessitates post-mapping computed tomography (CT) to confirm the locations of dye markings. We hypothesized that electromagnetic navigation bronchoscopy (ENB) simplifies VAL-MAP by omitting post-mapping CT. Under general anesthesia
Geoff Nelson et al.
Medical physics, 43(12), 6282-6282 (2016-12-03)
Electromagnetic navigation bronchoscopy (ENB) provides improved targeting accuracy during transbronchial biopsies of suspicious nodules. The greatest weakness of ENB-based guidance is the registration divergence that exists between the planning CT, acquired days or weeks before the intervention, and the patient
Imane Laraba et al.
PloS one, 16(1), e0245037-e0245037 (2021-01-13)
The Fusarium sambucinum species complex (FSAMSC) is one of the most taxonomically challenging groups of fusaria, comprising prominent mycotoxigenic plant pathogens and other species with various lifestyles. Among toxins produced by members of the FSAMSC, trichothecenes pose the most significant

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service