Skip to Content
Merck
All Photos(1)

Key Documents

115428

Sigma-Aldrich

2-Methyl-3-nitroanisole

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)OCH3
CAS Number:
Molecular Weight:
167.16
Beilstein:
2328599
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
99%
Pricing and availability is not currently available.

Quality Level

Assay

99%

form

solid

mp

54-56 °C (lit.)

SMILES string

COc1cccc(c1C)[N+]([O-])=O

InChI

1S/C8H9NO3/c1-6-7(9(10)11)4-3-5-8(6)12-2/h3-5H,1-2H3

InChI key

HQCZLEAGIOIIMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Methyl-3-nitroanisole may be used in chemical synthesis studies.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service