Skip to Content
Merck
All Photos(1)

Key Documents

109649

Sigma-Aldrich

2-Methylanthraquinone

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O2
CAS Number:
Molecular Weight:
222.24
Beilstein:
2050523
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

95%

impurities

3-4% 1-methylanthraquinone

bp

236-238 °C/10 mmHg (lit.)

mp

170-173 °C (lit.)

SMILES string

Cc1ccc2C(=O)c3ccccc3C(=O)c2c1

InChI

1S/C15H10O2/c1-9-6-7-12-13(8-9)15(17)11-5-3-2-4-10(11)14(12)16/h2-8H,1H3

InChI key

NJWGQARXZDRHCD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service