Skip to Content
Merck
All Photos(1)

Key Documents

108405

Sigma-Aldrich

Methyl dichloroacetate

≥99%

Synonym(s):

Dichloroacetic acid methyl ester, NSC 2882

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2CHCOOCH3
CAS Number:
Molecular Weight:
142.97
Beilstein:
1747542
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

≥99%

refractive index

n20/D 1.442 (lit.)

bp

143 °C (lit.)

mp

−52 °C (lit.)

density

1.381 g/mL at 25 °C (lit.)

SMILES string

COC(=O)C(Cl)Cl

InChI

1S/C3H4Cl2O2/c1-7-3(6)2(4)5/h2H,1H3

InChI key

HKMLRUAPIDAGIE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in:
  • Nucleophilic substitution reactions with nitro(pentafluorosulfanyl)benzenes
  • Azide-alkyl halide cycloaddition and atom transfer radical addition
  • Chemical modification of STn antigen for enhancement of synthetic carbohydrate vaccines
  • Three component reaction of arynes with cyclic ethers and active methines
  • Catalytic radical haloalkylation for synthesis of neodysidenin
  • Addition reactions for syntehsis of enantiopure 3,4-diamino esters or amides

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service