Merck
  • L-DOPA is incorporated into brain proteins of patients treated for Parkinson's disease, inducing toxicity in human neuroblastoma cells in vitro.
L-DOPA is incorporated into brain proteins of patients treated for Parkinson's disease, inducing toxicity in human neuroblastoma cells in vitro.
Experimental neurology (2011-10-18)
Sandra W Chan, Rachael A Dunlop, Anthony Rowe, Kay L Double, Kenneth J Rodgers
ABSTRACT
Levodopa (L-DOPA), a close structural analogue of the protein amino acid L-tyrosine, can substitute for L-tyrosine in protein synthesis and be mistakenly incorporated into newly synthesised proteins in vitro. We show that L-DOPA-containing proteins are present in the brain in L-DOPA-treated Parkinson's disease patients and accumulate in specific brain regions. In vitro studies demonstrate that substitution of L-tyrosine residues in proteins with L-DOPA causes protein misfolding and promotes protein aggregation in SH-SY5Y neuroblastoma cells resulting in the appearance of autofluorescent bodies. We show that the presence of L-DOPA-containing proteins causes profound changes in mitochondria and stimulates the formation of autophagic vacuoles in cells. Unlike L-DOPA, which is toxic to cells through its ability to generate radicals, proteins containing incorporated L-DOPA are toxic to SH-SY5Y cells by a mechanism independent of oxidative stress and resistant to antioxidants. These data suggest that the accumulation of L-DOPA-containing proteins in vulnerable cells might negatively impact on cell function.
Sigma-Aldrich
2,2′-Biquinoline-4,4′-dicarboxylic acid, ≥90% (TLC)