Skip to Content
Merck
All Photos(1)

Key Documents

V900673

Sigma-Aldrich

Sodium hydrogentartrate

Vetec, reagent grade, 98%

Synonym(s):

L(+)-Tartaric acid monosodium salt, Sodium bitartrate, Tartaric acid monosodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaO2CCH(OH)CH(OH)CO2H
CAS Number:
Molecular Weight:
172.07
Beilstein:
6099125
EC Number:
MDL number:
UNSPSC Code:
51113400
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

Assay

98%

mp

253 °C (dec.) (lit.)

SMILES string

[Na+].O[C@H]([C@@H](O)C([O-])=O)C(O)=O

InChI

1S/C4H6O6.Na/c5-1(3(7)8)2(6)4(9)10;/h1-2,5-6H,(H,7,8)(H,9,10);/q;+1/p-1/t1-,2-;/m1./s1

InChI key

NKAAEMMYHLFEFN-ZVGUSBNCSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service