Skip to Content
Merck
All Photos(1)

Key Documents

V800056

Sigma-Aldrich

Ammonium thiocyanate

AR, ≥99%

Synonym(s):

Ammonium rhodanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4SCN
CAS Number:
Molecular Weight:
76.12
Beilstein:
3595135
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
AR
form:
crystals

grade

AR

vapor pressure

0.000114 hPa ( 20 °C)

product line

Vetec

Assay

≥99%

form

crystals

pH

4.8-5.8 (5% in water)

mp

152-154 °C (lit.)

anion traces

chloride (Cl-): ≤0.005%
sulfate (SO42-): ≤0.0025%
sulfide (S2-): ≤0.002%

cation traces

Fe: ≤0.0001%
heavy metals (as Pb): ≤0.0004%

SMILES string

N.SC#N

InChI

1S/CHNS.H3N/c2-1-3;/h3H;1H3

InChI key

SOIFLUNRINLCBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kamlesh Shrivas et al.
Journal of hazardous materials, 161(1), 325-329 (2008-05-09)
A novel and sensitive spectrophotometric method for the determination of molybdenum at trace levels in environmental and biological samples is proposed. The method is based on the reaction of Mo (V) with thiocyanate (SCN(-)) and methyltrioctyl ammonium chloride (MTOAC) in
Anders Clemmensen et al.
Experimental dermatology, 18(11), 979-984 (2009-08-04)
To obtain a separation of the epidermal and dermal compartments to examine compartment specific biological mechanisms in the skin, we incubated 4 mm human skin punch biopsies in ammonium thiocyanate. We wanted to test (i) the histological quality of the
Xiaolian Gu et al.
The Journal of investigative dermatology, 135(8), 2077-2083 (2015-04-02)
Epigenetic modifications by DNA methylation are associated with a wide range of diseases. Previous studies in psoriasis have concentrated on epigenetic changes in immune cells or in total skin biopsies that include stromal-associated changes. In order to improve our understanding
G Almanzar et al.
Journal of immunological methods, 387(1-2), 36-42 (2012-10-02)
Antibody avidity, defined as the strength of binding between antibody and antigen, represents a functional measure of affinity maturation of antibodies. Determination of the antibody avidity is usually performed separating high and low avidity antibodies by dissociating agents, but measurement
Andrea Trost et al.
Experimental dermatology, 16(3), 185-190 (2007-02-09)
As global transcriptome analyses with a growing demand on layer-specific applications are widely used in cutaneous biology, we investigated the effect of established and optimized dermo-epidermal separation methods on the quality of RNA. We compared enzymatic separation with dispase, chemical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service