Skip to Content
Merck
All Photos(1)

Documents

V800050

Sigma-Aldrich

Ammonium molybdate tetrahydrate

AR, ≥99%

Synonym(s):

Ammonium heptamolybdate tetrahydrate, Molybdic acid ammonium salt tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)6Mo7O24 · 4H2O
CAS Number:
Molecular Weight:
1235.86
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

AR

product line

Vetec

Assay

≥99%

form

crystalline powder

pH

4.0-5.5 (0.05 M in water)

density

2.498 g/mL at 25 °C (lit.)

anion traces

AsO43-, PO43- and SiO2: ≤0.0005%
chloride (Cl-): ≤0.0002%
sulfate (SO42-): ≤0.02%

cation traces

Cu: ≤0.001%
Fe: ≤0.001%
heavy metals (as Pb): ≤0.001%

SMILES string

N.N.N.N.N.N.O.O.O.O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O[Mo](O)(=O)=O.O[Mo](O)(=O)=O.O[Mo](O)(=O)=O

InChI

1S/7Mo.6H3N.10H2O.18O/h;;;;;;;6*1H3;10*1H2;;;;;;;;;;;;;;;;;;/q;;;;3*+2;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/p-6

InChI key

FIXLYHHVMHXSCP-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chuanxin Hou et al.
Journal of colloid and interface science, 596, 396-407 (2021-04-14)
The agaric-like anodes of porous carbon decorated with MoO2 nanoparticles (MoO2/C) for reversible Li/Na storage were synthesized via a green and facile bio-inspired route. The uniformly distributed MoO2 nanoparticles, the porous agaric-like carbon matrix and high degree graphitization of carbon
Shifeng Cao et al.
International journal of food microbiology, 141(3), 173-176 (2010-07-08)
The potential enhancement of Pichia membranifaciens by ammonium molybdate (NH(4)Mo) to control blue mould caused by Penicillium expansum on peach fruit was investigated. Combining P. membranifaciens at 1x10(8) cell/ml with 1 mM NH(4)Mo provided a more effective control of blue
Pradeep Verma et al.
Bioresource technology, 102(4), 3941-3945 (2010-12-24)
Pretreatments for enzymatic saccharification are crucial for the establishment of lignocellulosic biorefineries. In this study, we focused on ammonium ions and peroxometal complexes as potential delignifying agents. We first examined the pretreatment of beech wood with nine different ammonium salts
Kyung Ho Park et al.
Journal of hazardous materials, 138(2), 311-316 (2006-07-25)
The petroleum refining industry makes extensive use of hydroprocessing catalysts. These catalysts contain environmentally critical and economically valuable metals such as Mo, V, Ni and Co. In the present study, a simple hydrometallurgical processing of spent hydrodesulphurization (HDS) catalyst for
Sidney Omelon et al.
Electrophoresis, 28(16), 2808-2811 (2007-07-21)
Nonradioactive polyphosphate (poly(P); (PO(3) (-))(n)) species resolved by PAGE can be detected by hydrolytic degradation of the polyphosphates into orthophosphates (P(i)) with a 5 M HCl solution saturated with NaCl, followed by staining the P(i) degradation products in a 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service