Skip to Content
Merck
All Photos(1)

Key Documents

TA9H9876F401

Targetmol Chemicals Inc.

NE-100 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
391.98

Iupac Name

N-{2-[4-methoxy-3-(2-phenylethoxy)phenyl]ethyl}-N-propyl-1-propanamine hydrochloride

Inchi Code

1S/C23H33NO2.ClH/c1-4-15-24(16-5-2)17-13-21-11-12-22(25-3)23(19-21)26-18-14-20-9-7-6-8-10-20;/h6-12,19H,4-5,13-18H2,1-3H3;1H

InChI key

ZHGMDXSHODHWHV-UHFFFAOYSA-N

storage temp.

-80/-20

Purity

98.85%

Country of Origin

CN

Shipping Temperature

4

Physical Form

solid

Product Link

https://www.targetmol.com/compound/NE100 HCl

MSDS

https://www.targetmol.com/compound/NE100 HCl


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library