Skip to Content
Merck
All Photos(1)

Documents

Z777310

Supelco

Zenix®-C SEC-150 (3 µm) HPLC Columns

L x I.D. 15 cm × 4.6 mm, diol bonding phase

Synonym(s):

Zenix®-C SEC-150 Gel Filtration Column, 3 μm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115709
NACRES:
SB.52

product name

Zenix®-C SEC-150 HPLC Column, 3 μm particle size, L × I.D. 15 cm × 4.6 mm

material

stainless steel hardware

Agency

suitable for USP L59

product line

Zenix®

mol wt

500-150,000

feature

endcapped: no

packaging

pkg of 1 unit

manufacturer/tradename

Sepax 233150-4615

parameter

20 Mpa max. pressure (3000 psi)
80 °C max. temp.

technique(s)

HPLC: suitable

L × I.D.

15 cm × 4.6 mm

impurities

<10 ppm metals

matrix

spherical silica particle platform

matrix active group

diol bonding phase

particle size

3 μm

pore size

150 Å pore size

operating pH range

2.0-8.5

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

The bonded phase in Zenix-C SEC columns is more polar than the hydrophilic phase used in Zenix columns, making Zenix-C columns better suited for the gel filtration analysis of more hydrophobic bioopolymers such as membrane proteins and pegylated antibodies. Both Zenix-C and Zenix columns are based on the same high efficiency 3 micron particles and each product line is available in the same wide range of pore sizes to tailor the optimal pore size to the hydrodynamic radius of the biopolymer of interest.

Zenix User Manual

Legal Information

Zenix is a registered trademark of Sepax Technologies

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service