G005794
HPLC Analysis of Insulin (Species Variants) on BIOshell A160 Peptide C18
application for HPLC
About This Item
technique(s)
HPLC: suitable
test parameters
column: BIOshell A160 Peptide C18, 15 cm x 2.1 mm I.D., 2.7 μm particles (66905-U)
mobile phase: [A] 70:30, water with 0.1% TFA:acetonitrile with 0.09% TFA; [B] 55:45, water with 0.1% TFA:acetonitrile with 0.09% TFA
gradient: 0 to 100% B in 7.5 min
flow rate: 0.3 mL/min
pressure: 4040 psi (278 bar)
column temp.: 30 °C
detector: UV, 215 nm
injection: 0.5 μL
sample: 200 μg/mL insulin (bovine, human, porcine) in 0.1% formic acid
suitability
application for HPLC
application(s)
life science and biopharma
pharmaceutical (small molecule)
Analysis Note
Legal Information
Analyte
analytical column
standard
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service