Skip to Content
Merck
All Photos(1)

Documents

814025

TSKgel® Ether-5PW (20 µm) HPLC Columns

L x I.D. 1 cm × 8 mm, ether phase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

product name

TSKgel® Ether-5PW HPLC Column, phase ether, L × I.D. 1 cm × 8 mm, 20 μm particle size, glass

material

glass

product line

TSKgel®

manufacturer/tradename

Tosoh 14025

parameter

≤4 M salt concentration
≤50% organic solvent
10-50 °C temperature

technique(s)

HPLC: suitable

L × I.D.

1 cm × 8 mm

matrix

hydroxylated methacrylic polymer

matrix active group

ether phase

particle size

20 μm

operating pH

2-12

separation technique

hydrophobic interaction (HIC)

Looking for similar products? Visit Product Comparison Guide

General description

Both hydrophobic interaction chromatography (HIC) and reversed-phase liquid chromatography separate on the basis of protein hydrophobicity and allow selective binding and desorption of proteins. However, HIC operates at significantly lower binding energy and uses aqueous mobile phases. These characteristics provide a gentle technique that is less likely to disturb protein conformation. As a result, HIC generally provides better activity recovery.
Three resin-based TSKgel HIC columns provide a range of hydrophobicities for chromatographic optimization. TSKgel Phenyl-5PW columns are most hydrophobic, followed by Ether-5PW, while the surface of Butyl-NPR columns is most polar or least hydrophobic. Ammonium sulfate concentration can be minimized by using a more hydrophobic packing such as Phenyl-5PW. We recommend the more polar Ether-5PW column for purifying more hydrophobic proteins. Ether-5PW and Phenyl-5PW packings are based on TSKgel G5000PW resin - 10 micron particles with 1000  pores. Butyl-NPR packing is prepared from 2.5 micron nonporous particles, allowing for rapid analyses and semi-micropreparative applications. All three column types can be cleaned with 0.2 M NaOH. Scale-up can be performed by using corresponding Toyopearl bulk resins.
Each TSKgel® Guardgel Kit contains 5 mL packing material, specified in the product description, in an appropriate slurry solvent. Guardgel Kits to protect glass columns also include a column holder, a glass column, 10 mm L x 13 mm OD x 8 mm ID, 2 external endfittings, 2 caps with M6 threads, and a packing adaptor. Simple instructions to pack the guard column are included in the shipment.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service