Skip to Content
Merck
All Photos(1)

Documents

43002

Supelco

all-cis-7,10,13,16,19-Docosapentaenoic acid

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H34O2
CAS Number:
Molecular Weight:
330.50
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.24

grade

analytical standard

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

storage temp.

−20°C

SMILES string

CC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CCCCCC(O)=O

InChI

1S/C22H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24/h3-4,6-7,9-10,12-13,15-16H,2,5,8,11,14,17-21H2,1H3,(H,23,24)/b4-3-,7-6-,10-9-,13-12-,16-15-

InChI key

YUFFSWGQGVEMMI-JLNKQSITSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service