30282-U
Replaceable fluoropolymer-faced silicone septa for ZHE Fitting
pkg of 10 ea
Synonym(s):
Septa, PTFE/Silicone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
PTFE face
septum (fluoropolymer-faced silicone)
silicone body
packaging
pkg of 10 ea
parameter
250 °C temp. limit
application(s)
environmental
industrial hygiene
compatibility
for use with Zero Headspace Extractor Fitting (ZHE)
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service