29851-U
SGE Syringes for eVol® XCHANGE® Analytical Syringe Digital Drive
volume 20-500 μL, needle size 23 ga, needle L × O.D. × I.D. 50 mm × 0.63 mm × 0.32 mm, bevel tip stainless steel needle, pkg of 1 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
bevel tip stainless steel needle
needle size
23 ga
feature
500 μL eVol® syringe
packaging
pkg of 1 ea
manufacturer/tradename
SGE 2910024
needle L × O.D. × I.D.
50 mm × 0.63 mm × 0.32 mm
volume
20-500 μL
Looking for similar products? Visit Product Comparison Guide
General description
XCHANGE® analytical syringes can be easily and quickly changed. This allows the user to choose the best syringe for the volume being measured. It also allows the user to dedicate individual syringe to specific liquids or methods, reducing the possibility of cross-contamination. Only a few XCHANGE syringes are requied to dispense liquid volumes from 0.2 μL to 500 μL.
Legal Information
XCHANGE is a registered trademark of SGE Analytical Science Pty Ltd.
eVol is a registered trademark of SGE Analytical Science Pty Ltd.
related product
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service