Skip to Content
Merck
All Photos(1)

Key Documents

24623

Supelco

Vacuum Sampling Pumps

Model 1062 Bag Sampler (Single 1-10 Liter Bags)

Synonym(s):

Air Sampling Pumps

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008

description

Not UL listed; Not EX rated

technique(s)

whole air sampling: suitable

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for use with Single 1-10 Liter Bags

Looking for similar products? Visit Product Comparison Guide

General description

Vacuum Sampling Pumps allows to capture a discrete air sample in a 10-liter bag without having the air sample pass through a vacuum pump first. It is a portable vacuum chamber-based sampler, which provides fast zero cross-contamination sampling of refinery stack gases and VOC vent gases.

Other Notes

The battery charger is not included with part number 24623. Please order part number 24644 separately.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service