Skip to Content
Merck
All Photos(1)

Documents

22114

Fine Metering Valve

brass, 1/8 in. angle arms

Synonym(s):

Swagelok® SS2-A,BRASS METERING VALVE 1/8A EA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115717
eCl@ss:
32035302

material

brass

description

fine adjustment

fitting

1/8 in. angle arms

Looking for similar products? Visit Product Comparison Guide

General description

Use for very accurate flow regulation. Install a flow measuring device downstream. Nuts and ferrules included. An optional vernier handle (Cat. No. 22122, order separately) is available for added convenience.

Application

Fine metering valve may be used on the exit of a flow system, which is basically used for the conductance measurements of decyl-, dodecyl-, tetradecyl- and hexadecyltrimethylammonium bromide in water at 25, 60, 95, 130, and 160°C, primarily done in order to determine their critical micelle concentrations (CMCs).

Legal Information

Swagelok is a registered trademark of Swagelok Company

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Critical micelle concentrations for alkyltrimethylammonium bromides in water from 25 to 160 C.
Fennell ED
Journal of Solution Chemistry, 13 (2), 87-101 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service