Documents
20480
Supelco
"Y" GlasSeal Connector, Borosilicate Glass
pkg of 1 ea
About This Item
UNSPSC Code:
41121800
packaging
pkg of 1 ea
technique(s)
gas chromatography (GC): suitable
General description
Column connectors are useful for attaching a guard column/retention gap to an analytical column, or for repairing a broken column.
Quickly connect three pieces of fused silica tubing of the same or different diameters without the need for tools. The leak-free connection is useful to split a sample to two columns, or to split a column effluent to two detectors. Silanized for an inert inside surface. For use with 0.10 - 0.53 mm I.D. fused silica tubing.
Other Notes
We offer a variety of chromatography accessories including analytical syringes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)