20050-U
ORBO™ 608 Amberlite™ XAD®-2 (20/50) 150/75 mg
W,W,W separators, O.D. × L 8 mm × 110 mm, pkg of 50 ea
Synonym(s):
ORBO™ Amberlite™ XAD-2 Tube
About This Item
Recommended Products
material
W,W,W separators
Agency
NIOSH 2514,2543,5514
product line
Amberlite™
ORBO™
composition
Bed A, 150 mg
Bed B, 75 mg
packaging
pkg of 50 ea
manufacturer/tradename
ORBO™ 608
technique(s)
active air sampling: suitable
O.D. × L
8 mm × 110 mm
matrix
XAD-2
particle size
20-50 mesh
application(s)
air monitoring
environmental
industrial hygiene
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service