150695
Perchloric acid solution
0.1 M in glacial acetic acid, for 3S adapter technology
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HClO4
CAS Number:
Molecular Weight:
100.46
MDL number:
UNSPSC Code:
12352301
Recommended Products
form
liquid
reaction suitability
reaction type: Acid-base reactions
concentration
0.0995-0.1005 M
0.1 M in glacial acetic acid
technique(s)
titration: suitable
volume
1 L
1000 mL
pH
0.1 (20 °C)
density
1.06 g/cm3 at 20 °C
SMILES string
OCl(=O)(=O)=O
InChI
1S/ClHO4/c2-1(3,4)5/h(H,2,3,4,5)
InChI key
VLTRZXGMWDSKGL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
3S - Safe, Smart, Secure
An intelligent packaging solution protected with originality seal.
Reagent information is transferred via RFID from reagent bottle to titrator.
3S reagents are compatible only with the Metrohm Omnis titration platform.
Please see here SigmaAldrich.com/3S
An intelligent packaging solution protected with originality seal.
Reagent information is transferred via RFID from reagent bottle to titrator.
3S reagents are compatible only with the Metrohm Omnis titration platform.
Please see here SigmaAldrich.com/3S
Other Notes
Once the bottle is opened, storage time is limited.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
104.0 °F
Flash Point(C)
40 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service