Skip to Content
Merck
All Photos(14)

Documents

ZRB012657

Sigma-Aldrich

Anti-PLA2R1 Antibody, clone 1D5, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

180 kDa secretory phospholipase A2 receptor, C-type lectin domain family 13-member C, M-type receptor., PLA2-R, Secretory phospholipase A2 receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1D5, recombinant monoclonal
monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 168.6 kDa
observed mol wt ~220 kDa

species reactivity

human, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable using 1:100
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable using 1:100
western blot: suitable using 1:1,000

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 1D5 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects human Secretory phospholipase A2 receptor (PLA2R1). It targets an epitope with in the extracellular domain.

Immunogen

GST/His-tagged recombinant fragment corresponding to 136 amino acids from the N-terminal, extracellular domain region of human Secretory phospholipase A2 receptor (PLA2R1).

Application

Anti-PLA2R1, clone 1D5, ZooMAb, Cat. No. ZRB012657, is a recombinant Rabbit Monoclonal antibody that detects Secretory phospholipase A2 receptor and has been tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected PLA2R1 in HeLa, A431, HUVEC and NIH 3T3 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected PLA2R1 in human kidney tissue sections.

Target description

Secretory phospholipase A2 receptor (UniProt: also known as PLA2-R, PLA2R, 180 kDa secretory phospholipase A2 receptor, C-type lectin domain family 13-member C, M-type receptor) is encoded by the PLA2R1 (also known as CLEC13C) gene (Gene ID: 22925) in human. PLA2R is a single-pass type I membrane protein that serves as a receptor for secretory phospholipase A2 (sPLA2). It is highly expressed in kidney and is also present in lung macrophages, pancreas, amnion, and placenta. PLA2R is a receptor for phospholipase sPLA2-IB/PLA2G1B, but not sPLA2-IIA/PLA2G2A. It can also bind to snake PA2-like toxins. Binding of sPLA2 to its receptor participates in both positive and negative regulation of sPLA2 functions and clearance. Binding of sPLA2-IB/PLA2G1B is shown to induce various effects depending on the cell type, such as activation of the mitogen-activated protein kinase cascade to induce cell proliferation, the production of lipid mediators, selective release of arachidonic acid in bone marrow-derived mast cells. PLA2R is also involved in responses in proinflammatory cytokine productions during endotoxic shock. The circulating, soluble form of secretory phospholipase A2 receptor acts as a negative regulator of sPLA2 functions by blocking the biological functions of sPLA2-IB/PLA2G1B. PLA2R is initially produced with a signal peptide sequence (aa 1-20), the cleavage of which produced the mature form that has a large extracellular domain (aa 21-1397), a transmembrane domain (aa 1398-1418), and a cytoplasmic tail (aa 1419-1463). The extracellular domain contains a fibronectin-like type II domain (173-221), and a tandem repeat of eight carbohydrate-recognition domains (C-type lectins). This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

120 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service