Documents
Z709611
Sartorius mLINE® mechanical pipettor
8 channel, volume 30-300 μL
Synonym(s):
Biohit pipettes, pipettor
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
feature
8-channel
electronic: no
packaging
pack of 1 ea
manufacturer/tradename
Sartorius 725140
volume measuring range
30-300 μL
volume
300 μL
General description
Sartorius′s new mechanical pipettor family - the mLINE - offers a unique combination of novel features for mechanical pipetting. Excellent performance, full autoclavability, smooth ergonomics and perfect safety. The mLINE pipettors cover the full volume range of 0.1 to 5000.
  • Extremely light pipetting and tip ejection
  • Unique safety features
  • Stepless volume adjustment and patented volume locking system
  • Thermal insulation of internal components to improve accuracy
  • Full autoclavability without disassembling
  • Easy cleaning, maintenance and calibration
  • ID tag and color-coded caps
  • Safe-Cone Filters and unique ejection
  • Opti-load tip mounting mechanism for even tip sealing and light tip ejection in multichannel pipettors
Legal Information
mLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy