Documents
Z708305
Slide mailer, 5 place
side opening
About This Item
UNSPSC Code:
41123302
NACRES:
MA.01
material
polypropylene
feature
Holds 5 x slides 76x26 mm
packaging
package of 25 Slide mailers
manufacturer/tradename
Heathrow Scientific HS15982
size
81 mm × 19 mm × 31 mm
General description
Ideal for shipping, storing or staining slides. These polypropylene containers hold 5 standard slides (3 × 1 in. or 25 × 75 mm). Slotted channels inside the mailer keep slides safely separated. Both end-opening and side-opening versions close securely to keep slides safe. The side-opening mailer measures 81 × 19 × 31 mm; end-opening mailer is 43 × 24 × 88 mm.