Z692670
microDOC Compact Gel Documentation System, with UV Transilluminator (UVT312)
with TotalLab™ 1D software, AC/DC input 110 V AC
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41105300
manufacturer/tradename
Cleaver CSL-MDOCUV3121D$
Looking for similar products? Visit Product Comparison Guide
General description
This option combines the microDOC Compact Gel Documentation System with a 21 x 21 cm tranilluminator.
Available with optional TotalLab™ 1D software. Main software functions include: lane creation, background subtraction, band detection, profile deconvolution, and molecular size, pI and quantity calibration.
System comprises of:
Camera: 1/1.7-inch type CMOS sensor with DIGIC 5 processor; 5x optical / 4x digital; 12.1 megapixels;
4GB storage card
Ethidium bromide & close up +3 filters
Lift-off hood with easy access door and built-in inner lights
Computer Interface: Hi-Speed USB (Mini-B compatible)
Screen: 8” TFT
21 x 21 cm transilluminator
Available with optional TotalLab™ 1D software. Main software functions include: lane creation, background subtraction, band detection, profile deconvolution, and molecular size, pI and quantity calibration.
System comprises of:
Camera: 1/1.7-inch type CMOS sensor with DIGIC 5 processor; 5x optical / 4x digital; 12.1 megapixels;
4GB storage card
Ethidium bromide & close up +3 filters
Lift-off hood with easy access door and built-in inner lights
Computer Interface: Hi-Speed USB (Mini-B compatible)
Screen: 8” TFT
21 x 21 cm transilluminator
Legal Information
TotalLab is a trademark of Nonlinear Dynamics Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service